3-(3-Chloro-4-fluorophenoxy)pyrrolidine hydrochloride
Supplier: Matrix Scientific
023385-500MG
101815-900EA
186.15
USD
101815-900
3-(3-Chloro-4-fluorophenoxy)pyrrolidine hydrochloride
3-(3-Chloro-4-fluorophenoxy)pyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₂Cl₂FNO |
MDL Number:
MFCD11506401 CAS Number: 1185025-66-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...