3-(3-Fluorophenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

023383-500MG
101815-896EA 315.38 USD
101815-896
3-(3-Fluorophenoxy)pyrrolidine hydrochloride
3-(3-Fluorophenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₃ClFNO
MDL Number: MFCD09879239
CAS Number: 1185119-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR