3-(3-Bromophenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

023382-500MG
101815-894EA 402.5 USD
101815-894
3-(3-Bromophenoxy)pyrrolidine hydrochloride
3-(3-Bromophenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₃BrClNO
MDL Number: MFCD09879238
CAS Number: 1185298-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR