3-(2-Ethylphenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

023380-500MG
101815-890EA 315.38 USD
101815-890
3-(2-Ethylphenoxy)pyrrolidine hydrochloride
3-(2-Ethylphenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₈ClNO
MDL Number: MFCD09879237
CAS Number: 1185299-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR