3-(2,5-Dimethylphenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

023378-500MG
101815-886EA 235.75 USD
101815-886
3-(2,5-Dimethylphenoxy)pyrrolidine hydrochloride
3-(2,5-Dimethylphenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₈ClNO
MDL Number: MFCD11506400
CAS Number: 1185301-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR