3-(5-Isopropyl-2-methylphenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

023377-500MG
101815-884EA 315.38 USD
101815-884
3-(5-Isopropyl-2-methylphenoxy)pyrrolidine hydrochloride
3-(5-Isopropyl-2-methylphenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₂ClNO
MDL Number: MFCD13561086
CAS Number: 1220033-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR