3-(2,3,5-Trimethylphenoxy)pyrrolidine

Supplier: Matrix Scientific

023375-500MG
101815-880EA 469.23 USD
101815-880
3-(2,3,5-Trimethylphenoxy)pyrrolidine
3-(2,3,5-Trimethylphenoxy)pyrrolidine

Hazard: Irritant

Formula: C₁₃H₁₉NO
MDL Number: MFCD08688565
CAS Number: 946726-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR