3-(2,3-Dimethylphenoxy)pyrrolidine

Supplier: Matrix Scientific

023374-500MG
101815-878EA 315.38 USD
101815-878
3-(2,3-Dimethylphenoxy)pyrrolidine
3-(2,3-Dimethylphenoxy)pyrrolidine

Hazard: Irritant

Formula: C₁₂H₁₇NO
MDL Number: MFCD08687663
CAS Number: 946715-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR