3-(2-Methylphenoxy)pyrrolidine

Supplier: Matrix Scientific

023373-500MG
101815-876EA 315.38 USD
101815-876
3-(2-Methylphenoxy)pyrrolidine
3-(2-Methylphenoxy)pyrrolidine

Hazard: Irritant

Formula: C₁₁H₁₅NO
MDL Number: MFCD08060984
CAS Number: 46196-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR