3-[2-(Trifluoromethyl)phenoxy]pyrrolidine hydrochloride

Supplier: Matrix Scientific

023371-500MG
101815-872EA 315.38 USD
101815-872
3-[2-(Trifluoromethyl)phenoxy]pyrrolidine hydrochloride
3-[2-(Trifluoromethyl)phenoxy]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₃ClF₃NO
MDL Number: MFCD11100505
CAS Number: 1185301-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR