3-([1,1'-Biphenyl]-2-yloxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

023370-500MG
101815-870EA 495.38 USD
101815-870
3-([1,1'-Biphenyl]-2-yloxy)pyrrolidine hydrochloride
3-([1,1'-Biphenyl]-2-yloxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₁₈ClNO
MDL Number: MFCD09879235
CAS Number: 1185002-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR