3-(2-Ethoxyphenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

023369-500MG
101815-868EA 495.38 USD
101815-868
3-(2-Ethoxyphenoxy)pyrrolidine hydrochloride
3-(2-Ethoxyphenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₈ClNO₂
MDL Number: MFCD09879234
CAS Number: 1185301-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR