3-(2-Methoxyphenoxy)pyrrolidine

Supplier: Matrix Scientific

023368-500MG
101815-866EA 315.38 USD
101815-866
3-(2-Methoxyphenoxy)pyrrolidine
3-(2-Methoxyphenoxy)pyrrolidine

Hazard: Irritant

Formula: C₁₁H₁₅NO₂
MDL Number: MFCD08060986
CAS Number: 21767-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR