3-(2,5-Dichlorophenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

023366-500MG
101815-862EA 315.38 USD
101815-862
3-(2,5-Dichlorophenoxy)pyrrolidine hydrochloride
3-(2,5-Dichlorophenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₂Cl₃NO
MDL Number: MFCD26142751
CAS Number: 1707365-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR