3-(2-Chlorophenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

023362-500MG
101815-854EA 235.75 USD
101815-854
3-(2-Chlorophenoxy)pyrrolidine hydrochloride
3-(2-Chlorophenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₃Cl₂NO
MDL Number: MFCD09879232
CAS Number: 1185298-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR