3-Phenoxypyrrolidine hydrochloride

Supplier: Matrix Scientific

023359-500MG
101815-848EA 429.23 USD
101815-848
3-Phenoxypyrrolidine hydrochloride
3-Phenoxypyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₄ClNO
MDL Number: MFCD09879231
CAS Number: 21767-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR