3-(Tetrahydro-2-furanylmethoxy)pyrrolidine

Supplier: Matrix Scientific

023356-500MG
101815-842EA 315.38 USD
101815-842
3-(Tetrahydro-2-furanylmethoxy)pyrrolidine
3-(Tetrahydro-2-furanylmethoxy)pyrrolidine

Hazard: Irritant

Formula: C₉H₁₇NO₂
MDL Number: MFCD08688542
CAS Number: 933702-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR