3-[(2-Methyl-2-propenyl)oxy]pyrrolidine hydrochloride
Supplier: Matrix Scientific
023355-500MG
101815-840EA
315.38
USD
101815-840
3-[(2-Methyl-2-propenyl)oxy]pyrrolidine hydrochloride
3-[(2-Methyl-2-propenyl)oxy]pyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₈H₁₆ClNO |
MDL Number:
MFCD11100504 CAS Number: 1185297-81-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...