3-([1,1'-Biphenyl]-4-ylmethoxy)pyrrolidine
Supplier: Matrix Scientific
023352-500MG
101815-834EA
315.38
USD
101815-834
3-([1,1'-Biphenyl]-4-ylmethoxy)pyrrolidine
3-([1,1'-Biphenyl]-4-ylmethoxy)pyrrolidine
Hazard: Irritant
Formula:
C₁₇H₁₉NO |
MDL Number:
MFCD08686923 CAS Number: 946726-60-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...