3-[(2-Methylbenzyl)oxy]pyrrolidine hydrochloride

Supplier: Matrix Scientific

023347-500MG
101815-824EA 495.38 USD
101815-824
3-[(2-Methylbenzyl)oxy]pyrrolidine hydrochloride
3-[(2-Methylbenzyl)oxy]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₈ClNO
MDL Number: MFCD11100502
CAS Number: 1185297-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR