3-[(2-Methoxybenzyl)oxy]pyrrolidine hydrochloride

Supplier: Matrix Scientific

023346-500MG
101815-822EA 315.38 USD
101815-822
3-[(2-Methoxybenzyl)oxy]pyrrolidine hydrochloride
3-[(2-Methoxybenzyl)oxy]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₈ClNO₂
MDL Number: MFCD12405048
CAS Number: 1185303-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR