3-(Benzyloxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

023345-500MG
101815-820EA 244.95 USD
101815-820
3-(Benzyloxy)pyrrolidine hydrochloride
3-(Benzyloxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₆ClNO
MDL Number: MFCD11845784
CAS Number: 127342-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR