3-Ethoxypyrrolidine hydrochloride

Supplier: Matrix Scientific

023343-500MG
101815-816EA 315.38 USD
101815-816
3-Ethoxypyrrolidine hydrochloride
3-Ethoxypyrrolidine hydrochloride

Hazard: Irritant

Formula: C₆H₁₄ClNO
MDL Number: MFCD09879229
CAS Number: 1185299-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR