3-Propoxypyrrolidine

Supplier: Matrix Scientific

023340-500MG
101815-810EA 235.75 USD
101815-810
3-Propoxypyrrolidine
3-Propoxypyrrolidine

Hazard: Irritant

Formula: C₇H₁₅NO
MDL Number: MFCD08688520
CAS Number: 946681-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR