3-(Cyclopropylmethoxy)pyrrolidine

Supplier: Matrix Scientific

023332-500MG
101815-794EA 402.5 USD
101815-794
3-(Cyclopropylmethoxy)pyrrolidine
3-(Cyclopropylmethoxy)pyrrolidine

Hazard: Irritant

Formula: C₈H₁₅NO
MDL Number: MFCD08687524
CAS Number: 932702-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR