4-(2,4-Dichloro-3,5-dimethylphenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

023326-500MG
101815-782EA 315.38 USD
101815-782
4-(2,4-Dichloro-3,5-dimethylphenoxy)piperidine hydrochloride
4-(2,4-Dichloro-3,5-dimethylphenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈Cl₃NO
MDL Number: MFCD12405064
CAS Number: 1185297-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR