4-(2-Methoxy-5-nitrophenoxy)piperidine

Supplier: Matrix Scientific

023323-500MG
101815-776EA 315.38 USD
101815-776
4-(2-Methoxy-5-nitrophenoxy)piperidine
4-(2-Methoxy-5-nitrophenoxy)piperidine

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₄
MDL Number: MFCD08688063
CAS Number: 946759-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR