4-(4-Bromo-2-fluorophenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

023322-500MG
101815-774EA 538.46 USD
101815-774
4-(4-Bromo-2-fluorophenoxy)piperidine hydrochloride
4-(4-Bromo-2-fluorophenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₄BrClFNO
MDL Number: MFCD11100501
CAS Number: 1185105-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR