4-(2,4-Difluorophenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

023321-500MG
101815-772EA 538.46 USD
101815-772
4-(2,4-Difluorophenoxy)piperidine hydrochloride
4-(2,4-Difluorophenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₄ClF₂NO
MDL Number: MFCD09997065
CAS Number: 1171775-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR