8-(4-Piperidinyloxy)quinoline dihydrochloride

Supplier: Matrix Scientific

023317-500MG
101815-764EA 495.38 USD
101815-764
8-(4-Piperidinyloxy)quinoline dihydrochloride
8-(4-Piperidinyloxy)quinoline dihydrochloride

Hazard: Irritant

Formula: C₁₄H₁₈Cl₂N₂O
MDL Number: MFCD09879222
CAS Number: 1185297-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR