6-Methyl-3-pyridinyl-4-piperidinyl ether dihydrochloride

Supplier: Matrix Scientific

023316-500MG
101815-762EA 1692.31 USD
101815-762
6-Methyl-3-pyridinyl-4-piperidinyl ether dihydrochloride
6-Methyl-3-pyridinyl-4-piperidinyl ether dihydrochloride

Hazard: Irritant

Formula: C₁₁H₁₈Cl₂N₂O
MW: 265.18 g/mol
MDL Number: MFCD11845781
CAS Number: 1184991-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR