2-Naphthyl-4-piperidinyl ether

Supplier: Matrix Scientific

023313-500MG
101815-756EA 495.38 USD
101815-756
2-Naphthyl-4-piperidinyl ether
2-Naphthyl-4-piperidinyl ether

Hazard: Irritant

Formula: C₁₅H₁₇NO
MDL Number: MFCD08688441
CAS Number: 78055-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR