Methyl-2-[4-(4-piperidinyloxy)phenyl]acetate hydrochloride

Supplier: Matrix Scientific

023309-500MG
101815-750EA 384.62 USD
101815-750
Methyl-2-[4-(4-piperidinyloxy)phenyl]acetate hydrochloride
Methyl-2-[4-(4-piperidinyloxy)phenyl]acetate hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₀ClNO₃
MDL Number: MFCD10687221
CAS Number: 811813-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR