4-(4-Methylphenoxy)piperidine

Supplier: Matrix Scientific

023307-500MG
101815-748EA 315.38 USD
101815-748
4-(4-Methylphenoxy)piperidine
4-(4-Methylphenoxy)piperidine

Hazard: Irritant

Formula: C₁₂H₁₇NO
MDL Number: MFCD04115017
CAS Number: 63843-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR