4-[4-(sec-Butyl)phenoxy]piperidine
Supplier: Matrix Scientific
023306-500MG
101815-746EA
315.38
USD
101815-746
4-[4-(sec-Butyl)phenoxy]piperidine
4-[4-(sec-Butyl)phenoxy]piperidine
Hazard: Irritant
Formula:
C₁₅H₂₃NO |
MDL Number:
MFCD08687204 CAS Number: 946759-80-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...