4-(4-Isopropylphenoxy)piperidine hydrochloride
Supplier: Matrix Scientific
023305-500MG
101815-744EA
186.15
USD
101815-744
4-(4-Isopropylphenoxy)piperidine hydrochloride
4-(4-Isopropylphenoxy)piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₂ClNO |
MDL Number:
MFCD09879221 CAS Number: 1173079-45-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...