4-[4-(tert-Butyl)phenoxy]piperidine hydrochloride

Supplier: Matrix Scientific

023304-500MG
101815-742EA 402.5 USD
101815-742
4-[4-(tert-Butyl)phenoxy]piperidine hydrochloride
4-[4-(tert-Butyl)phenoxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO
MDL Number: MFCD11100500
CAS Number: 1184979-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR