4-[4-(Trifluoromethyl)phenoxy]piperidine hydrochloride

Supplier: Matrix Scientific

Synonyms: 4-(4-(Trifluoromethyl)phenoxy)piperidine hydrochloride

023302-500MG
101815-738EA 76.92 USD
101815-738
4-[4-(Trifluoromethyl)phenoxy]piperidine hydrochloride
4-[4-(Trifluoromethyl)phenoxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₅ClF₃NO₂
MW: 281.7000 g/mol
MDL Number: MFCD02258937
CAS Number: 287952-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR