(4-Chlorophenyl)[4-(4-piperidinyloxy)phenyl]methanone

Supplier: Matrix Scientific

023301-500MG
101815-736EA 315.38 USD
101815-736
(4-Chlorophenyl)[4-(4-piperidinyloxy)phenyl]methanone
(4-Chlorophenyl)[4-(4-piperidinyloxy)phenyl]methanone

Hazard: Irritant

Formula: C₁₈H₁₈ClNO₂
MDL Number: MFCD08687461
CAS Number: 946714-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR