4-([1,1'-Biphenyl]-4-yloxy)piperidine

Supplier: Matrix Scientific

023300-500MG
101815-734EA 315.38 USD
101815-734
4-([1,1'-Biphenyl]-4-yloxy)piperidine
4-([1,1'-Biphenyl]-4-yloxy)piperidine

Hazard: Irritant

Formula: C₁₇H₁₉NO
MDL Number: MFCD06246000
CAS Number: 397278-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR