4-(4-Ethoxyphenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

023299-500MG
101815-732EA 538.46 USD
101815-732
4-(4-Ethoxyphenoxy)piperidine hydrochloride
4-(4-Ethoxyphenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO₂
MDL Number: MFCD11845778
CAS Number: 1185303-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR