4-(4-Chloro-3,5-dimethylphenoxy)piperidine
Supplier: Matrix Scientific
023295-500MG
101815-724EA
315.38
USD
101815-724
4-(4-Chloro-3,5-dimethylphenoxy)piperidine
4-(4-Chloro-3,5-dimethylphenoxy)piperidine
Hazard: Irritant
Formula:
C₁₃H₁₈ClNO |
MDL Number:
MFCD08687285 CAS Number: 946726-36-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...