4-(4-Bromophenoxy)piperidine

Supplier: Matrix Scientific

023294-500MG
101815-722EA 235.75 USD
101815-722
4-(4-Bromophenoxy)piperidine
4-(4-Bromophenoxy)piperidine

Hazard: Irritant

Formula: C₁₁H₁₄BrNO
MDL Number: MFCD04115016
CAS Number: 74130-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR