3-Ethylphenyl-4-piperidinyl ether hydrochloride
Supplier: Matrix Scientific
023293-500MG
101815-720EA
315.38
USD
101815-720
3-Ethylphenyl-4-piperidinyl ether hydrochloride
3-Ethylphenyl-4-piperidinyl ether hydrochloride
Hazard: Irritant
Formula:
C₁₃H₂₀ClNO |
MDL Number:
MFCD11100499 CAS Number: 1338974-43-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...