4-(3-Methoxyphenoxy)piperidine

Supplier: Matrix Scientific

023287-500MG
101815-708EA 235.75 USD
101815-708
4-(3-Methoxyphenoxy)piperidine
4-(3-Methoxyphenoxy)piperidine

Hazard: Irritant

Formula: C₁₂H₁₇NO₂
MDL Number: MFCD08061092
CAS Number: 162402-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR