N-[3-(4-Piperidinyloxy)phenyl]acetamide hydrochloride

Supplier: Matrix Scientific

023286-500MG
101815-706EA 235.75 USD
101815-706
N-[3-(4-Piperidinyloxy)phenyl]acetamide hydrochloride
N-[3-(4-Piperidinyloxy)phenyl]acetamide hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O₂
MDL Number: MFCD09879220
CAS Number: 459819-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR