3-Iodophenyl-4-piperidinyl ether

Supplier: Matrix Scientific

023285-500MG
101815-704EA 315.38 USD
101815-704
3-Iodophenyl-4-piperidinyl ether
3-Iodophenyl-4-piperidinyl ether

Hazard: Irritant

Formula: C₁₁H₁₄INO
MDL Number: MFCD08687944
CAS Number: 946726-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR