4-(3-Bromophenoxy)piperidine hydrochloride
Supplier: Matrix Scientific
023281-500MG
101815-698EA
315.38
USD
101815-698
4-(3-Bromophenoxy)piperidine hydrochloride
4-(3-Bromophenoxy)piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₅BrClNO |
MDL Number:
MFCD09879219 CAS Number: 1072944-50-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...