2-Allylphenyl-4-piperidinyl ether

Supplier: Matrix Scientific

023280-500MG
101815-696EA 284.62 USD
101815-696
2-Allylphenyl-4-piperidinyl ether
2-Allylphenyl-4-piperidinyl ether

Hazard: Irritant

Formula: C₁₄H₁₉NO
MDL Number: MFCD08686775
CAS Number: 946714-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR