2-Ethylphenyl-4-piperidinyl ether hydrochloride

Supplier: Matrix Scientific

023279-500MG
101815-694EA 495.38 USD
101815-694
2-Ethylphenyl-4-piperidinyl ether hydrochloride
2-Ethylphenyl-4-piperidinyl ether hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO
MDL Number: MFCD11845775
CAS Number: 1185301-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR